Learn More
Human Cardiotrophin-1/CT-1 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB2602-100
 View more versions of this product
Product Code. 18741137
  • £415.00 / 100µg
Description
Cardiotrophin-1/CT-1 Monoclonal specifically detects Cardiotrophin-1/CT-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cardiotrophin-1/CT-1
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
cardiophin 1, cardiotrophin 1, cardiotrophin-1, Cardiotrophin1, CT-1CT1, CTF1
E. coli-derived recombinant human Cardiotrophin-1 Ser2-Ala201 Accession # Q16619
100 μg
Primary
Detects human Cardiotrophin-1 in direct ELISAs and Western blots.
Human
IgG2a
Western Blot, ELISA, Immunohistochemistry
89221
Western Blot 2 ug/mL, ELISA, Immunohistochemistry 5-25 ug/mL
Q16619
Mouse
Protein A or G purified from ascites
RUO
1489
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.