missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Carboxypeptidase A2/CPA2 Antibody, R&D Systems™

Goat Polyclonal Antibody
£146.00 - £415.00
Specifications
| Antigen | Carboxypeptidase A2/CPA2 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18799237
![]() |
R&D Systems
AF2896-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18072412
|
R&D Systems
AF2896 |
100 μg |
£415.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carboxypeptidase A2/CPA2 Polyclonal specifically detects Carboxypeptidase A2/CPA2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Carboxypeptidase A2/CPA2 | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| 1358 | |
| NS0-derived rhCPA2 (aa 17 - 417) aa 17-419 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human CPA2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human (rh) CPA1 is observed and less than 1% cross-reactivity with rhCPB1 and recombinant mouse CPA4 is observed. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| carboxypeptidase A2, carboxypeptidase A2 (pancreatic), CPA2, EC 3.4.17, EC 3.4.17.15 | |
| CPA2 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title