missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Carboxypeptidase A2/CPA2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2896
This item is not returnable.
View return policy
Description
Carboxypeptidase A2/CPA2 Polyclonal specifically detects Carboxypeptidase A2/CPA2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Carboxypeptidase A2/CPA2 | |
| Polyclonal | |
| Unconjugated | |
| carboxypeptidase A2, carboxypeptidase A2 (pancreatic), CPA2, EC 3.4.17, EC 3.4.17.15 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1358 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| CPA2 | |
| NS0-derived rhCPA2 (aa 17 - 417) aa 17-419 | |
| 100 μg | |
| Primary | |
| Detects human CPA2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human (rh) CPA1 is observed and less than 1% cross-reactivity with rhCPB1 and recombinant mouse CPA4 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction