missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human C5L2/GPR77 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £412.00
Specifications
| Antigen | C5L2/GPR77 |
|---|---|
| Clone | 650918 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18742177
![]() |
R&D Systems
MAB10254-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18763737
![]() |
R&D Systems
MAB10254-100 |
100 μg |
£412.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C5L2/GPR77 Monoclonal specifically detects C5L2/GPR77 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| C5L2/GPR77 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| C5a anaphylatoxin chemotactic receptor C5L2, C5L2G-protein coupled receptor 77, G protein-coupled receptor 77, G protein-coupled receptor C5L2, GPF77, GPR77 | |
| NS0 mouse myeloma cell line transfected with human C5L2/GPR77, Met1-Val337, Accession # Q9P296 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human C5L2/GPR77 in direct ELISAs. |
| 650918 | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9P296 | |
| 27202 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title