missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human C5L2/GPR77 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10254-SP
This item is not returnable.
View return policy
Description
C5L2/GPR77 Monoclonal specifically detects C5L2/GPR77 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| C5L2/GPR77 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| C5a anaphylatoxin chemotactic receptor C5L2, C5L2G-protein coupled receptor 77, G protein-coupled receptor 77, G protein-coupled receptor C5L2, GPF77, GPR77 | |
| NS0 mouse myeloma cell line transfected with human C5L2/GPR77, Met1-Val337, Accession # Q9P296 | |
| 25 μg | |
| Primary | |
| Detects human C5L2/GPR77 in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 650918 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q9P296 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27202 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction