missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human BMP-7 Recombinant Protein
Recombinant Human BMP7 is a bioactive protein intended for use in cell culture applications
£220.00 - £8700.00
Specifications
For Use With (Application) | Cell Culture |
---|---|
Molecular Weight (g/mol) | 38 kDa |
Purification Method | Gel-Purified |
Gene Alias | BMP-7 |
Protein Length | 139 aa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11360852
|
Gibco™
PHC9544 |
10μg |
£220.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11340852
|
Gibco™
PHC9541 |
100μg |
£2135.00
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11350852
|
Gibco™
PHC9543 |
1mg |
£8700.00
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Human BMP-7 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
BMP7 is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta (TGF-β) superfamily. The TGF-β superfamily includes many families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo at an extraskeletal site.- High purity: minimal interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to help ensure a biologically active protein
- High biological activity: in-house activity testing includes a variety of cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays; specific activity is determined by the dose dependent induction of alkaline phosphatase production in the ATDC-5 cell line (Mouse chondrogenic cell line)
Shipping Condition: Dry Ice.
Specifications
Cell Culture | |
Gel-Purified | |
139 aa | |
Store in freezer (-5 to -30°C). | |
50 ng/ml | |
< 0.5 EU/μg | |
Mammalian | |
Recombinant, Ligand | |
BMPs (Bone Morphogenetic Proteins) & Related Factors | |
Gibco™ |
38 kDa | |
BMP-7 | |
Human | |
Carrier-Free | |
Induction of alkaline phosphatase in ATDC5 cells | |
Dry Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Bone Research, Stem Cell Research, Differentiation | |
95 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.