missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human BMP-7 Recombinant Protein

Product Code. 11340852
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11340852 100 μg Each
11350852 1 mg Each
11360852 10 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11340852 Supplier Gibco™ Supplier No. PHC9541

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

Carrier-Free Reconstitution: Centrifuge the vial briefly, before opening to bring the contents to the bottom. Reconstitute the lyophilized protein in sterile water containing 0.1% human serum albumin. Apportion the reconstituted protein into working aliquots and store at = -20°C. Make any further dilutions of the reconstituted protein in low endotoxin medium or a buffered solution containing a carrier protein such as heat inactivated FCS or tissue culture grade BSA. Storage: Store the lyophilized protein at = -20°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at = -20°C. Avoid repeated freeze-thaw cycles.

The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extra skeletal site. Based on its expression early in embryogenesis, BMP7 has a proposed role in early development. In addition, the fact that BMP7 is closely related to BMP5 and BMP6 has lead to speculation of possible bone inductive activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P18075, P23359
For Use With (Application) Bioactivity
Formulation 10mM acetic acid with no preservative
Gene ID (Entrez) 12162, 655
Molecular Weight (g/mol) 38 kDa
Name Human BMP-7
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <0.5 EU/μg
Gene Alias BMP; BMP7; BMP-7; bone moorphogenic protein-7; Bone morphogenetic protein; bone morphogenetic protein 7; bone morphogenetic protein 7 (osteogenic protein 1); bone morphogenetic protein 7 preproprotein; bone morphogenic protein 7; Eptotermin alfa; OP1; OP-1; Osteogenic protein 1; RP11-560A15.5
Common Name BMP-7
Gene Symbol Bmp7
Biological Activity ED50 = 8.31 ng/mL; determined by the dose-dependent induction of alkaline phosphatase production in ATDC-5 cells.
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Content And Storage -20°C
Classification Carrier-Free
Activity ED50 = 8.31 ng/mL; determined by the dose-dependent induction of alkaline phosphatase production in ATDC-5 cells.
Bioassay Induction of alkaline phosphatase in ATDC5 cells
Endotoxin Level <0.5 EU/μg
Shipping Condition Dry Ice
Expression System HEK293 cells
Protein Family Growth Factors & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype BMPs (Bone Morphogenetic Proteins) & Related Factors
Research Category Bone Research, Stem Cell Research, Differentiation
Product Line Gibco
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.