missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BMP-5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£156.00 - £474.00
Specifications
| Antigen | BMP-5 |
|---|---|
| Clone | 78205 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18748428
![]() |
R&D Systems
MAB715-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18183411
|
R&D Systems
MAB715 |
500 μg |
£474.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BMP-5 Monoclonal specifically detects BMP-5 in Human samples. It is validated for Western Blot.Specifications
| BMP-5 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP5, BMP-5, bone morphogenetic protein 5, MGC34244 | |
| BMP5 | |
| IgG2a | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 78205 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P22003 | |
| 653 | |
| Mouse myeloma cell line NS0-derived recombinant human BMP-5 Accession # P22003 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human BMP-5 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title