missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BMP-5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB715-SP
This item is not returnable.
View return policy
Description
BMP-5 Monoclonal specifically detects BMP-5 in Human samples. It is validated for Western Blot.
Specifications
| BMP-5 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP5, BMP-5, bone morphogenetic protein 5, MGC34244 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 653 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 78205 | |
| Western Blot 1 ug/mL | |
| P22003 | |
| BMP5 | |
| Mouse myeloma cell line NS0-derived recombinant human BMP-5 Accession # P22003 | |
| 25 μg | |
| Primary | |
| Detects human BMP-5 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction