missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BMP-2/BMP-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£168.00 - £1775.00
Specifications
| Antigen | BMP-2/BMP-4 |
|---|---|
| Clone | 65529 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18768309
![]() |
R&D Systems
MAB355-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18757799
![]() |
R&D Systems
MAB355-100 |
100 μg |
£451.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18732827
![]() |
R&D Systems
MAB355-500 |
500 μg |
£1775.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BMP-2/BMP-4 Monoclonal specifically detects BMP-2/BMP-4 in Human samples. It is validated for Western Blot.Specifications
| BMP-2/BMP-4 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P12643 | |
| 650 | |
| E. coli-derived recombinant human BMP-2 Ala284-Arg396 Accession # P12643 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human BMP-2 and human BMP-4 in direct ELISAs and Western blots. In direct ELISAs, this antibody shows no cross-reactivity with recombinant human (rh) BMP-5 or rhBMP-6. |
| 65529 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BDA2, BMP2, BMP-2, BMP2A, BMP-2A, BMP2ABone morphogenetic protein 2A, BMP2B, BMP2B1, BMP4, bone morphogenetic protein 2, MCOPS6, OFC11, SSFSC | |
| BMP2 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title