missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BMP-2/BMP-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB355-100
This item is not returnable.
View return policy
Description
BMP-2/BMP-4 Monoclonal specifically detects BMP-2/BMP-4 in Human samples. It is validated for Western Blot.
Specifications
| BMP-2/BMP-4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BDA2, BMP2, BMP-2, BMP2A, BMP-2A, BMP2ABone morphogenetic protein 2A, BMP2B, BMP2B1, BMP4, bone morphogenetic protein 2, MCOPS6, OFC11, SSFSC | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 650 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 65529 | |
| Western Blot 1 ug/mL | |
| P12643 | |
| BMP2 | |
| E. coli-derived recombinant human BMP-2 Ala284-Arg396 Accession # P12643 | |
| 100 μg | |
| Primary | |
| Detects human BMP-2 and human BMP-4 in direct ELISAs and Western blots. In direct ELISAs, this antibody shows no cross-reactivity with recombinant human (rh) BMP-5 or rhBMP-6. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction