missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £378.00
Specifications
| Antigen | ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase |
|---|---|
| Clone | 511702 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18795739
![]() |
R&D Systems
MAB4494-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18112551
![]() |
R&D Systems
MAB4494 |
100 μg |
£378.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase Monoclonal specifically detects ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q02083 | |
| 27163 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ASAHL/N-acylethanolamine-hydrolyzing Acid A isoform 1 Ser29-Lys359 (Val151Ile) Accession # Q02083 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human ASAHL/N-acylethanolamine-hydrolyzing Acid A in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse ASAHL is observed and no cross-reactivity with recombinant human ASAH2 or recombinant mouse ASAH2 is observed. |
| 511702 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Acid ceramidase-like protein, ASAHL, ASAH-like protein, EC 3.5.1.-, NAAA, N-acylethanolamine acid amidase, N-acylethanolamine-hydrolyzing acid amidase, Nacylethanolaminehydrolyzing Acid Amidase, N-acylsphingosine amidohydrolase (acid ceramidase)-like, N-acylsphingosine amidohydrolase-like, PLT | |
| NAAA | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title