missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4494
This item is not returnable.
View return policy
Description
ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase Monoclonal specifically detects ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q02083 | |
| NAAA | |
| Chinese hamster ovary cell line CHO-derived recombinant human ASAHL/N-acylethanolamine-hydrolyzing Acid A isoform 1 Ser29-Lys359 (Val151Ile) Accession # Q02083 | |
| 100 μg | |
| Primary | |
| Detects human ASAHL/N-acylethanolamine-hydrolyzing Acid A in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse ASAHL is observed and no cross-reactivity with recombinant human ASAH2 or recombinant mouse ASAH2 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 511702 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Acid ceramidase-like protein, ASAHL, ASAH-like protein, EC 3.5.1.-, NAAA, N-acylethanolamine acid amidase, N-acylethanolamine-hydrolyzing acid amidase, Nacylethanolaminehydrolyzing Acid Amidase, N-acylsphingosine amidohydrolase (acid ceramidase)-like, N-acylsphingosine amidohydrolase-like, PLT | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27163 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction