missing translation for 'onlineSavingsMsg'
Learn More

Human Aquaporin 4/AQP4 Antibody, Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

£206.00 - £476.00

Specifications

Antigen Aquaporin-4
Clone 1093722
Dilution Immunohistochemistry 3-25 μg/mL, Multiplex Immunofluorescence 25 μg/mL
Applications Multiplex Immunofluorescence, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30306119
View Documents
R&D Systems
MAB11659-100
100 μg
£476.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30306105
View Documents
R&D Systems
MAB11659-SP
25 μg
£206.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Aquaporin-4 Monoclonal antibody specifically detects Aquaporin-4 in Human samples. It is validated for Multiplex Immunofluorescence,Immunohistochemistry
Specifications

Specifications

Aquaporin-4
Immunohistochemistry 3-25 μg/mL, Multiplex Immunofluorescence 25 μg/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2μm filtered solution in PBS.
AQP4, AQP-4, aquaporin 4, MGC22454, MIWC, WCH4
Synthetic Peptide, Accession # P55087
Primary
Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Antibody
1093722
Multiplex Immunofluorescence, Immunohistochemistry
Unconjugated
Mouse
Human
P55087
361
IgG
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.