Learn More
Human Aquaporin 4/AQP4 Antibody, Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB11659-SP
 View more versions of this product
Product Code. 30306105
  • £206.00 / 25µg
Description
Aquaporin-4 Monoclonal antibody specifically detects Aquaporin-4 in Human samples. It is validated for Multiplex Immunofluorescence,Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Aquaporin-4
Monoclonal
Unconjugated
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2μm filtered solution in PBS.
AQP4, AQP-4, aquaporin 4, MGC22454, MIWC, WCH4
Synthetic Peptide, Accession # P55087
25 μg
Primary
Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Purified
Multiplex Immunofluorescence, Immunohistochemistry
1093722
Immunohistochemistry 3-25 μg/mL, Multiplex Immunofluorescence 25 μg/mL
P55087
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
361