missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Apolipoprotein E3/ApoE3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £420.00
Specifications
| Antigen | Apolipoprotein E3/ApoE3 |
|---|---|
| Clone | 377108 |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18773747
![]() |
R&D Systems
MAB41444-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18721617
![]() |
R&D Systems
MAB41444-100 |
100 μg |
£420.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Apolipoprotein E3/ApoE3 Monoclonal specifically detects Apolipoprotein E3/ApoE3 in Human samples. It is validated for ELISA.Specifications
| Apolipoprotein E3/ApoE3 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P02649 | |
| 348 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 377108 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| AD2, Apo-E, APOE, ApoE4, apolipoprotein E, LDLCQ5, LPG | |
| E. coli-derived human Apolipoprotein E3/ApoE3 protein, Lys19-His317, Accession # P02649 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title