missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Apolipoprotein E3/ApoE3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB41444-100
This item is not returnable.
View return policy
Description
Apolipoprotein E3/ApoE3 Monoclonal specifically detects Apolipoprotein E3/ApoE3 in Human samples. It is validated for ELISA.
Specifications
| Apolipoprotein E3/ApoE3 | |
| Monoclonal | |
| Unconjugated | |
| P02649 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 348 | |
| Human | |
| Purified |
| ELISA | |
| 377108 | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| AD2, Apo-E, APOE, ApoE4, apolipoprotein E, LDLCQ5, LPG | |
| E. coli-derived human Apolipoprotein E3/ApoE3 protein, Lys19-His317, Accession # P02649 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction