missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Aminopeptidase O/ONPEP Antibody, R&D Systems™

Goat Polyclonal Antibody
£167.00 - £417.00
Specifications
| Antigen | Aminopeptidase O/ONPEP |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18717977
![]() |
R&D Systems
AF4647-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18053102
![]() |
R&D Systems
AF4647 |
100 μg |
£417.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aminopeptidase O/ONPEP Polyclonal specifically detects Aminopeptidase O/ONPEP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
| Aminopeptidase O/ONPEP | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| aminopeptidase O, AOPEP, APO, AP-OFLJ55832, C90RF3, C9orf3, chromosome 9 open reading frame 3, EC 3.4.11.-, FLJ14675, FLJ40923, ONPEP | |
| C9ORF3 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Q8N6M6 | |
| 84909 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Aminopeptidase O/ONPEP isoform 1 Met1-Phe819 Accession # Q8N6M6 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Aminopeptidase O/ONPEP in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title