missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Aminopeptidase O/ONPEP Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4647
This item is not returnable.
View return policy
Description
Aminopeptidase O/ONPEP Polyclonal specifically detects Aminopeptidase O/ONPEP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| Aminopeptidase O/ONPEP | |
| Polyclonal | |
| Unconjugated | |
| Q8N6M6 | |
| C9ORF3 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Aminopeptidase O/ONPEP isoform 1 Met1-Phe819 Accession # Q8N6M6 | |
| 100 μg | |
| Primary | |
| Detects human Aminopeptidase O/ONPEP in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
| aminopeptidase O, AOPEP, APO, AP-OFLJ55832, C90RF3, C9orf3, chromosome 9 open reading frame 3, EC 3.4.11.-, FLJ14675, FLJ40923, ONPEP | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 84909 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction