missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human alpha-Smooth Muscle Actin Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
£256.00 - £451.00
Specifications
| Antigen | alpha-Smooth Muscle Actin |
|---|---|
| Clone | 1A4 |
| Concentration | 0.2 mg/mL |
| Dilution | Intracellular Staining by Flow Cytometry 5 uL/10^6 cells |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18145081
|
R&D Systems
IC1420V-100 |
100 Tests |
£451.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18700027
![]() |
R&D Systems
IC1420V-025 |
25 Tests |
£256.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
alpha-Smooth Muscle Actin Monoclonal specifically detects alpha-Smooth Muscle Actin in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| alpha-Smooth Muscle Actin | |
| 0.2 mg/mL | |
| Flow Cytometry | |
| Alexa Fluor 405 | |
| Mouse | |
| Human | |
| AAT6, ACTA2, Actin alpha 2, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, alphaSmooth Muscle Actin, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, MYMY5, sma, smooth muscle actin | |
| ACTA2 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human alpha-Smooth Muscle Actin in flow cytometry. |
| 1A4 | |
| Intracellular Staining by Flow Cytometry 5 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 59 | |
| alpha-Smooth Muscle Actin N-terminal Synthetic Decapeptide | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title