Learn More
Human alpha-Smooth Muscle Actin Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems IC1420V-025
 View more versions of this product
Product Code. 18700027
  • £256.00 / 25 tests
Description
alpha-Smooth Muscle Actin Monoclonal specifically detects alpha-Smooth Muscle Actin in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
alpha-Smooth Muscle Actin
Monoclonal
0.2 mg/mL
Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
AAT6, ACTA2, Actin alpha 2, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, alphaSmooth Muscle Actin, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, MYMY5, sma, smooth muscle actin
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
59
Human
Purified
Flow Cytometry
1A4
Alexa Fluor 405
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
ACTA2
alpha-Smooth Muscle Actin N-terminal Synthetic Decapeptide
25 Tests
Primary
Detects human alpha-Smooth Muscle Actin in flow cytometry.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG2a