missing translation for 'onlineSavingsMsg'
Learn More

Human alpha-Galactosidase A/GLA Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£167.00 - £421.00

Specifications

Antigen alpha-Galactosidase A/GLA
Clone 1029734
Dilution Immunocytochemistry 8-25 ug/mL
Applications Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18732437
GREENER_CHOICE
View Documents
R&D Systems
MAB61461-SP
25 μg
£167.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18764167
GREENER_CHOICE
View Documents
R&D Systems
MAB61461-100
100 μg
£421.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

alpha-Galactosidase A/GLA Monoclonal antibody specifically detects alpha-Galactosidase A/GLA in Human samples. It is validated for Immunocytochemistry
Specifications

Specifications

alpha-Galactosidase A/GLA
Immunocytochemistry 8-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Agalsidase, agalsidase alfa, Agalsidase alpha, Alpha-D-galactosidase A, Alpha-D-galactoside galactohydrolase, alpha-D-galactoside galactohydrolase 1, alpha-gal A, alpha-galactosidase A, EC 3.2.1, EC 3.2.1.22, GALA, galactosidase, alpha, melibiase
Chinese Hamster Ovary cell line CHO-derived human alpha-Galactosidase A/GLA protein, Met1-Leu429, Accession # P06280
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
1029734
Immunocytochemistry
Unconjugated
Mouse
Human
P06280
2717
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.