missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human alpha-Galactosidase A/GLA Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB61461-SP
This item is not returnable.
View return policy
Description
alpha-Galactosidase A/GLA Monoclonal antibody specifically detects alpha-Galactosidase A/GLA in Human samples. It is validated for Immunocytochemistry
Specifications
| alpha-Galactosidase A/GLA | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Agalsidase, agalsidase alfa, Agalsidase alpha, Alpha-D-galactosidase A, Alpha-D-galactoside galactohydrolase, alpha-D-galactoside galactohydrolase 1, alpha-gal A, alpha-galactosidase A, EC 3.2.1, EC 3.2.1.22, GALA, galactosidase, alpha, melibiase | |
| Chinese Hamster Ovary cell line CHO-derived human alpha-Galactosidase A/GLA protein, Met1-Leu429, Accession # P06280 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Immunocytochemistry | |
| 1029734 | |
| Immunocytochemistry 8-25 ug/mL | |
| P06280 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2717 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction