missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human AIM2 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£178.00 - £370.00
Specifications
| Antigen | AIM2 |
|---|---|
| Clone | 2307A |
| Dilution | Western Blot 2 μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18018141
![]() |
R&D Systems
MAB9965-SP |
25 μg |
£178.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18058181
![]() |
R&D Systems
MAB9965-100 |
100 μg |
£370.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AIM2 Monoclonal specifically detects AIM2 in Human samples. It is validated for Western Blot.Specifications
| AIM2 | |
| Western Blot 2 μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| O14862 | |
| 9447 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. |
| 2307A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| absent in melanoma 2PYHIN4, Gm1313, Ifi210, interferon-inducible protein AIM2, PYHIN4 | |
| E. coli-derived recombinant human AIM2 Lys79-Thr180 Accession # O14862 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human AIM2 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title