missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human AIM2 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB9965-SP
This item is not returnable.
View return policy
Description
AIM2 Monoclonal specifically detects AIM2 in Human samples. It is validated for Western Blot.
Specifications
| AIM2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| absent in melanoma 2PYHIN4, Gm1313, Ifi210, interferon-inducible protein AIM2, PYHIN4 | |
| E. coli-derived recombinant human AIM2 Lys79-Thr180 Accession # O14862 | |
| 25 μg | |
| 9447 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
| IgG |
| Western Blot | |
| 2307A | |
| Western Blot 2 μg/mL | |
| O14862 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human AIM2 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction