missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAMTS13 Antibody, R&D Systems™

Sheep Polyclonal Antibody
£171.00 - £502.00
Specifications
| Antigen | ADAMTS13 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18777457
![]() |
R&D Systems
AF4245-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18040892
|
R&D Systems
AF4245 |
100 μg |
£502.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ADAMTS13 Polyclonal specifically detects ADAMTS13 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| ADAMTS13 | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Sheep | |
| Human | |
| A disintegrin and metalloproteinase with thrombospondin motifs 13, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 13, ADAM metallopeptidase with thrombospondin type 1 motif, 13, ADAM-TS13, ADAMTS-13, DKFZp434C2322, EC 3.4.24.14, EC 3.4.24.82, EC 3.4.24.87, FLJ42993, MGC118899, MGC118900, TTP, TTPADAM-TS 13, vWF-cleaving protease, vWF-CP, vWF-CPC9orf8, VWFCPvon Willebrand factor-cleaving protease | |
| ADAMTS13 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Q76LX8 | |
| 11093 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ADAMTS13 Gln34-Trp688 Accession # Q76LX8 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human ADAMTS13 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) ADAMTS14 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title