missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAMTS13 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF4245
This item is not returnable.
View return policy
Description
ADAMTS13 Polyclonal specifically detects ADAMTS13 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| ADAMTS13 | |
| Polyclonal | |
| Unconjugated | |
| Q76LX8 | |
| ADAMTS13 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ADAMTS13 Gln34-Trp688 Accession # Q76LX8 | |
| 100 μg | |
| Primary | |
| Detects human ADAMTS13 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) ADAMTS14 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| A disintegrin and metalloproteinase with thrombospondin motifs 13, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 13, ADAM metallopeptidase with thrombospondin type 1 motif, 13, ADAM-TS13, ADAMTS-13, DKFZp434C2322, EC 3.4.24.14, EC 3.4.24.82, EC 3.4.24.87, FLJ42993, MGC118899, MGC118900, TTP, TTPADAM-TS 13, vWF-cleaving protease, vWF-CP, vWF-CPC9orf8, VWFCPvon Willebrand factor-cleaving protease | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 11093 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction