missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAM8 aa 158-497 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£171.00 - £1775.00
Specifications
| Antigen | ADAM8 |
|---|---|
| Clone | 143303 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18753577
![]() |
R&D Systems
MAB1031-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18741919
![]() |
R&D Systems
MAB1031-100 |
100 μg |
£436.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18791197
![]() |
R&D Systems
MAB1031-500 |
500 μg |
£1775.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ADAM8 Monoclonal specifically detects ADAM8 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| ADAM8 | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| a disintegrin and metalloproteinase domain 8, ADAM 8, ADAM metallopeptidase domain 8, CD156a, CD156a antigen, CD156human leukocyte differentiation antigen, Cell surface antigen MS2, EC 3.4.24, EC 3.4.24.-, MGC134985, MS2, MS2disintegrin and metalloproteinase domain-containing protein 8 | |
| ADAM8 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 143303 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P78325 | |
| 101 | |
| Mouse myeloma cell line NS0-derived recombinant human ADAM8 ectodomain Asp158-Ser653 Accession # P78325 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human ADAM8 in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with the ectodomains of recombinant human ADAM9, 10, 15, 17, and 19 is observed. Detects an epitope within aa 201-497 of recombinant human ADAM8 which corresponds to the metalloprotease and disintegrin-like domains. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title