missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAM8 aa 158-497 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1031-SP
This item is not returnable.
View return policy
Description
ADAM8 Monoclonal specifically detects ADAM8 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| ADAM8 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P78325 | |
| ADAM8 | |
| Mouse myeloma cell line NS0-derived recombinant human ADAM8 ectodomain Asp158-Ser653 Accession # P78325 | |
| 25 μg | |
| Primary | |
| Detects human ADAM8 in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with the ectodomains of recombinant human ADAM9, 10, 15, 17, and 19 is observed. Detects an epitope within aa 201-497 of recombinant human ADAM8 which corresponds to the metalloprotease and disintegrin-like domains. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 143303 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| a disintegrin and metalloproteinase domain 8, ADAM 8, ADAM metallopeptidase domain 8, CD156a, CD156a antigen, CD156human leukocyte differentiation antigen, Cell surface antigen MS2, EC 3.4.24, EC 3.4.24.-, MGC134985, MS2, MS2disintegrin and metalloproteinase domain-containing protein 8 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 101 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction