missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAM22 Antibody, R&D Systems™

Sheep Polyclonal Antibody
£168.00 - £432.00
Specifications
| Antigen | ADAM22 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18719267
![]() |
R&D Systems
AF4908-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18099051
![]() |
R&D Systems
AF4908 |
100 μg |
£432.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ADAM22 Polyclonal specifically detects ADAM22 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| ADAM22 | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Sheep | |
| Human | |
| ADAM 22, ADAM metallopeptidase domain 22, MDC2, metalloproteinase-disintegrin ADAM22-3, metalloproteinase-like, disintegrin-like, and cysteine-rich protein 2, MGC149832 | |
| ADAM22 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Q9P0K1 | |
| 53616 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ADAM22 Gly26-His528 (Leu78Val) Accession # Q9P0K1 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human ADAM22 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) ADAM8, rhADAM12, and rhADAM33 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title