missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAM22 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4908
This item is not returnable.
View return policy
Description
ADAM22 Polyclonal specifically detects ADAM22 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| ADAM22 | |
| Polyclonal | |
| Unconjugated | |
| Q9P0K1 | |
| ADAM22 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ADAM22 Gly26-His528 (Leu78Val) Accession # Q9P0K1 | |
| 100 μg | |
| Primary | |
| Detects human ADAM22 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) ADAM8, rhADAM12, and rhADAM33 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| ADAM 22, ADAM metallopeptidase domain 22, MDC2, metalloproteinase-disintegrin ADAM22-3, metalloproteinase-like, disintegrin-like, and cysteine-rich protein 2, MGC149832 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 53616 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction