missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAM12 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB44161-SP
This item is not returnable.
View return policy
Description
ADAM12 Monoclonal specifically detects ADAM12 in Human samples. It is validated for Immunoprecipitation.
Specifications
| ADAM12 | |
| Monoclonal | |
| LYOPH | |
| Immunoprecipitation 25 ug/mL | |
| O43184 | |
| ADAM12 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ADAM12 Arg29-Ser513 Accession # O43184 | |
| 25 μg | |
| Primary | |
| Detects human ADAM12 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human ADAM8, 9, 10, 15, 17, 19, 22, 23, 32, or 33 is observed. | |
| Human | |
| Purified |
| Immunoprecipitation | |
| 632525 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| a disintegrin and metalloproteinase domain 12 (meltrin alpha), ADAM 12, ADAM metallopeptidase domain 12, disintegrin and metalloproteinase domain-containing protein 12, EC 3.4.24, EC 3.4.24.58, MCMP, MCMPMltna, Meltrin alpha, Meltrin-alpha, MLTNA, MLTNEC 3.4.24.- | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8038 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction