missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAM12 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£171.00 - £380.00
Specifications
| Antigen | ADAM12 |
|---|---|
| Clone | 632525 |
| Concentration | LYOPH |
| Dilution | Immunoprecipitation 25 ug/mL |
| Applications | Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18777568
![]() |
R&D Systems
MAB44161-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18109210
![]() |
R&D Systems
MAB44161 |
100 μg |
£380.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ADAM12 Monoclonal specifically detects ADAM12 in Human samples. It is validated for Immunoprecipitation.Specifications
| ADAM12 | |
| LYOPH | |
| Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| O43184 | |
| 8038 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ADAM12 Arg29-Ser513 Accession # O43184 | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Detects human ADAM12 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human ADAM8, 9, 10, 15, 17, 19, 22, 23, 32, or 33 is observed. |
| 632525 | |
| Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| a disintegrin and metalloproteinase domain 12 (meltrin alpha), ADAM 12, ADAM metallopeptidase domain 12, disintegrin and metalloproteinase domain-containing protein 12, EC 3.4.24, EC 3.4.24.58, MCMP, MCMPMltna, Meltrin alpha, Meltrin-alpha, MLTNA, MLTNEC 3.4.24.- | |
| ADAM12 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title