missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Activin A Precursor Antibody, R&D Systems™

Mouse Monoclonal Antibody
£157.00 - £458.00
Specifications
| Antigen | Activin A |
|---|---|
| Clone | 132815 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18771539
![]() |
R&D Systems
MAB1203-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18196051
![]() |
R&D Systems
MAB1203 |
500 μg |
£458.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Activin A Monoclonal specifically detects Activin A in Human samples. It is validated for Western Blot.Specifications
| Activin A | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| INHBA | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 132815 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DNA construct encoding human Activin A pro region fused to mature human Activin beta B chain | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Activin A Precursor in direct ELISAs and Western blots. Recognizes an epitope found in the human Activin A pro region that is removed when the protein is processed to its mature form. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title