missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Activin A Precursor Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1203
This item is not returnable.
View return policy
Description
Activin A Monoclonal specifically detects Activin A in Human samples. It is validated for Western Blot.
Specifications
| Activin A | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| INHBA | |
| DNA construct encoding human Activin A pro region fused to mature human Activin beta B chain | |
| 500 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 132815 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects human Activin A Precursor in direct ELISAs and Western blots. Recognizes an epitope found in the human Activin A pro region that is removed when the protein is processed to its mature form. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction