missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human 20S Proteasome Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £352.00
Specifications
| Antigen | 20SProteasome |
|---|---|
| Clone | 863425 |
| Dilution | Immunohistochemistry 5-25μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18138910
![]() |
R&D Systems
MAB95541-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18113241
![]() |
R&D Systems
MAB95541-100 |
100 μg |
£352.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
20S Proteasome Monoclonal specifically detects 20S Proteasome in Human samples. It is validated for Immunohistochemistry.Specifications
| 20SProteasome | |
| Immunohistochemistry 5-25μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Human 20S Proteasome purified from human erthrocytes | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human 20S Proteasome in direct ELISAs. |
| 863425 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title