missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human 20S Proteasome Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB95541-100
This item is not returnable.
View return policy
Description
20S Proteasome Monoclonal specifically detects 20S Proteasome in Human samples. It is validated for Immunohistochemistry.
Specifications
| 20SProteasome | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| Human 20S Proteasome purified from human erthrocytes | |
| 100 μg | |
| Detects human 20S Proteasome in direct ELISAs. | |
| Human | |
| Supernatant |
| Immunohistochemistry | |
| 863425 | |
| Immunohistochemistry 5-25μg/mL | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction