missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HuC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£171.00 - £517.00
Specifications
| Antigen | HuC |
|---|---|
| Dilution | Western Blot 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18175783
|
Novus Biologicals
NBP2-29864-0.025MG |
0.025 mg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18735543
|
Novus Biologicals
NBP2-29864 |
£517.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
HuC Polyclonal specifically detects HuC in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| HuC | |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| RUO | |
| Mouse, Human | |
| Q14576 | |
| 1995 | |
| Synthetic peptide, amino acids 20-46 of HuC. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| Polyclonal | |
| Rabbit | |
| Neuroscience | |
| 50% saturated ammonium sulfate and PBS with No Preservative | |
| DKFZp547J036, ELAV (embryonic lethal, abnormal vision, Drosophila)-like 3 (Hu antigen C), ELAV-like protein 3, Hu antigen C, Hu-antigen C, HuC, HUCL, HUCParaneoplastic cerebellar degeneration-associated antigen, MGC20653, PLE21Paraneoplastic limbic encephalitis antigen 21 | |
| ELAVL3 | |
| IgG | |
| Affinity Purified | |
| Recognizes HuC. The molecular weight of the protein is 38,862 Daltons. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title