missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HuC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-29864-0.025MG
This item is not returnable.
View return policy
Description
HuC Polyclonal specifically detects HuC in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| HuC | |
| Polyclonal | |
| Western Blot 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| Q14576 | |
| ELAVL3 | |
| Synthetic peptide, amino acids 20-46 of HuC. | |
| 0.025 mg | |
| Neuroscience | |
| 1995 | |
| Mouse, Human | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| 50% saturated ammonium sulfate and PBS with No Preservative | |
| DKFZp547J036, ELAV (embryonic lethal, abnormal vision, Drosophila)-like 3 (Hu antigen C), ELAV-like protein 3, Hu antigen C, Hu-antigen C, HuC, HUCL, HUCParaneoplastic cerebellar degeneration-associated antigen, MGC20653, PLE21Paraneoplastic limbic encephalitis antigen 21 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Recognizes HuC. The molecular weight of the protein is 38,862 Daltons. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction