missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ HTMT dimaleate
GREENER_CHOICE

Product Code. 18055852
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18055852

Brand: Tocris Bioscience™ 0646/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

H1/ H2 agonist

H1 and H2 receptor agonist. 4x104 times more active than histamine in H2-mediated effects in natural suppressor cells. Increases intracellular Ca2+ and IP3 in lymphocytes through a binding site other than H1, H2 or H3.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 195867-54-0
Molecular Formula C27H37F3N4O9

Specifications

Quantity 50 mg
Formula Weight 614.57
Purity >99%
Molecular Weight (g/mol) 618.607
Chemical Name or Material HTMT dimaleate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.