accessibility menu, dialog, popup

UserName

CAS RN 195867-54-0

CH3ONHFFFHNNHNH2COHOH2COHOOOHOOH

CAS RN 195867-54-0

IUPAC Name: 6-{[2-(1H-imidazol-5-yl)ethyl]amino}-N-[4-(trifluoromethyl)phenyl]heptanamide; bis(formic acid); bis(prop-2-enoic acid)
Synonyms: bis(acrylic acid); bis(formic acid); histamine trifluoromethyl toluidide
Molecular Weight (g/mol): 618.61
Molecular Formula: C27H37F3N4O9
InChi Key: KIMGHGOBUFFPNE-UHFFFAOYNA-N
SMILES: OC=O.OC=O.OC(=O)C=C.OC(=O)C=C.CC(CCCCC(=O)NC1=CC=C(C=C1)C(F)(F)F)NCCC1=CN=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results