missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ HTMT dimaleate

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
H1 and H2 receptor agonist. 4x104 times more active than histamine in H2-mediated effects in natural suppressor cells. Increases intracellular Ca2+ and IP3 in lymphocytes through a binding site other than H1, H2 or H3.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 614.57 |
| Purity | >99% |
| Molecular Weight (g/mol) | 618.607 |
| Chemical Name or Material | HTMT dimaleate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction