missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPC111 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56344
This item is not returnable.
View return policy
Description
HSPC111 Polyclonal specifically detects HSPC111 in Human samples. It is validated for Western Blot.
Specifications
| HSPC111 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| HBV pre-S2 trans-regulated protein 3, HSPC111HSPC185, LOC51491, NOP15, NOP16 nucleolar protein homolog (yeast), nucleolar protein 16, nucleolar protein 16 homolog, nucleolar protein 16 homolog (yeast) | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9Y3C1 | |
| NOP16 | |
| Synthetic peptides corresponding to HSPC111(hypothetical protein HSPC111) The peptide sequence was selected from the middle region of HSPC111. Peptide sequence RKVKAMEVDIEERPKELVRKPYVLNDLEAEASLPEKKGNTLSRDLIDYVR. | |
| 100 μL | |
| Stem Cell Markers | |
| 51491 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction