Learn More
HSPB8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15287-1-AP-20UL
 View more versions of this product
Product Code. 16854404
  • £135.00 / 20µL
Description
The protein encoded by this gene belongs to the superfamily of small heat-shock proteins containing a conservative alpha-crystallin domain at the C-terminal part of the molecule. The expression of this gene in induced by estrogen in estrogen receptor-positive breast cancer cells, and this protein also functions as a chaperone in association with Bag3, a stimulator of macroautophagy. Thus, this gene appears to be involved in regulation of cell proliferation, apoptosis, and carcinogenesis, and mutations in this gene have been associated with different neuromuscular diseases, including Charcot-Marie-Tooth disease.
Specifications
Specifications
HSPB8
Polyclonal
Unconjugated
HSPB8
Alpha crystallin C chain, CMT2L, CRYAC, DHMN2, E2 induced gene 1 protein, E2IG1, H11, heat shock 22kDa protein 8, Heat shock protein beta 8, HMN2, HMN2A, HSP22, HSPB8, Protein kinase H11
Rabbit
Antigen Affinity Chromatography
RUO
113906, 26353, 80888
-20°C
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9EPX0, Q9JK92, Q9UJY1
Hspb8
HSPB8 Fusion Protein Ag7456
20 μL
Primary
Rat, Mouse, Human