missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSPB3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£155.00 - £386.00
Specifications
| Antigen | HSPB3 |
|---|---|
| Concentration | 0.66 mg/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17723294
|
Invitrogen™
PA5121894 |
20 μL |
£155.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17713294
![]() |
Invitrogen™
PA5121893 |
100 μL |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Inhibitor of actin polymerization.Specifications
| HSPB3 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q12988 | |
| 8988 | |
| Met1-Lys150 (Accession Q12988), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| HSPB3 |
| 0.66 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| DHMN2C; heat shock 17 kDa protein; heat shock 27kD protein 3; heat shock 27kD protein family, member 3; heat shock 27kDa protein 3; Heat shock protein; heat shock protein 3; heat shock protein B3; heat shock protein beta-3; heat shock protein family B (small) member 3; HMN2C; HSP; HSP 17; HSP27; HspB3; HSPL27; Protein 3 | |
| HSPB3 | |
| IgG | |
| Protein A, Antigen affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title