missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HSPB3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5121894

 View more versions of this product

Product Code. 17723294

  • £155.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Inhibitor of actin polymerization.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HSPB3
Polyclonal
Unconjugated
HSPB3
DHMN2C; heat shock 17 kDa protein; heat shock 27kD protein 3; heat shock 27kD protein family, member 3; heat shock 27kDa protein 3; Heat shock protein; heat shock protein 3; heat shock protein B3; heat shock protein beta-3; heat shock protein family B (small) member 3; HMN2C; HSP; HSP 17; HSP27; HspB3; HSPL27; Protein 3
Rabbit
Protein A, Antigen affinity chromatography
RUO
8988
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.66 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q12988
HSPB3
Met1-Lys150 (Accession Q12988), with N-terminal His Tag.
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.