missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA8/HSC71/Hsc70 Antibody (13D3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 19 publications
£274.00
Specifications
| Antigen | HSPA8/HSC71/Hsc70 |
|---|---|
| Clone | 13D3 |
| Dilution | Western Blot 1:1000 - 1:5000, Simple Western reported by internal validation, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunoprecipitation 2 ul, Immunohistochemistry-Paraffin 1:20 - 1:200, Immunohistochemistry-Frozen 1:20 - 1:200, Immunoblotting, Block/Neutralize |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
HSPA8/HSC71/Hsc70 Monoclonal specifically detects HSPA8/HSC71/Hsc70 in Human, Mouse, Rat, Bovine, Feline, Fish, Hamster, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Block/Neutralize.Specifications
| HSPA8/HSC71/Hsc70 | |
| Western Blot 1:1000 - 1:5000, Simple Western reported by internal validation, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunoprecipitation 2 ul, Immunohistochemistry-Paraffin 1:20 - 1:200, Immunohistochemistry-Frozen 1:20 - 1:200, Immunoblotting, Block/Neutralize | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| Heat shock 70 kDa protein 8, heat shock 70kD protein 8, heat shock 70kDa protein 8, heat shock cognate 71 kDa protein, heat shock cognate protein 54, heat shock cognate protein, 71-kDa, HSC54, HSC70heat shock 70kd protein 10, HSC71, HSP71, HSP73constitutive heat shock protein 70, HSPA10, LAP1, lipopolysaccharide-associated protein 1, LPS-associated protein 1, MGC131511, MGC29929, NIP71, N-myristoyltransferase inhibitor protein 71 | |
| HSPA8 | |
| IgM | |
| Unpurified | |
| This displays slight cross-reactivity to HSP 70. |
| 13D3 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Chaperone Mediated Autophagy (CMA) | |
| P08107 | |
| 3312 | |
| Mouse spermatogenic cell protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| 70 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title