missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSPA14 Recombinant Rabbit Monoclonal Antibody (23GB6565), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | HSPA14 |
|---|---|
| Clone | 23GB6565 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087497
![]() |
Invitrogen™
MA553066 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088151
|
Invitrogen™
MA553067 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSPA14 Recombinant Monoclonal antibody specifically detects HSPA14 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| HSPA14 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q0VDF9, Q6AYB4, Q99M31 | |
| 307133, 50497, 51182 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6565 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 70kDa; heat shock 70 kDa protein 14; heat shock 70kDa protein 14; heat shock 70kDa protein 14 isoform 1 variant 3; Heat shock protein; heat shock protein 14; heat shock protein 4; heat shock protein family A (Hsp70) member 14; heat shock protein HSP60; heat shock protein hsp70-related protein; heat shock protein, 70 kDa 4; HSP; HSP60; Hsp70-4; HSP70L1; HSP70-like protein 1; HSP7E; HSPA14; hsr.1; NST-1 | |
| A synthesized peptide derived from human HSPA14 (50-101AA). | |
| Primary | |
| -20°C | |
| HSPA14 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title