missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSPA14 Recombinant Rabbit Monoclonal Antibody (23GB6565), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553067
This item is not returnable.
View return policy
Description
The heat shock proteins (HSPs) comprise a group of highly conserved, abundantly expressed proteins with diverse functions, including the assembly and sequestering of multi-protein complexes, the transportation of nascent poly-peptide chains across cellular membranes and the regulation of protein folding. HSPA14 (heat shock 70kDa protein 14), also known as HSP70-4 or HSP70L1, is a 509 amino acid novel HSP protein derived from human dendritic cells. Belonging to the heat shock protein 70 family, HSPA14 is thought to promote dendritic cell maturation. It is also suggested that HSPA14 stimulates secretion of the proinflammatory cytokines interleukin 12p70 (IL-12p70), IL-1 beta, TNF alpha, and the chemokines IP-10, MIP-1 alpha, MIP-1 beta and normal T cell expressed and secreted (RANTES).
Specifications
| HSPA14 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| HSPA14 | |
| 70kDa; heat shock 70 kDa protein 14; heat shock 70kDa protein 14; heat shock 70kDa protein 14 isoform 1 variant 3; Heat shock protein; heat shock protein 14; heat shock protein 4; heat shock protein family A (Hsp70) member 14; heat shock protein HSP60; heat shock protein hsp70-related protein; heat shock protein, 70 kDa 4; HSP; HSP60; Hsp70-4; HSP70L1; HSP70-like protein 1; HSP7E; HSPA14; hsr.1; NST-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 307133, 50497, 51182 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6565 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q0VDF9, Q6AYB4, Q99M31 | |
| HSPA14 | |
| A synthesized peptide derived from human HSPA14 (50-101AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction