missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSPA12A Recombinant Rabbit Monoclonal Antibody (23GB6030), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | HSPA12A |
|---|---|
| Clone | 23GB6030 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089665
![]() |
Invitrogen™
MA552974 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088365
|
Invitrogen™
MA552975 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSPA12A Recombinant Monoclonal antibody specifically detects HSPA12A in Human, Mouse samples. It is validated for Western BlotSpecifications
| HSPA12A | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| O43301, Q8K0U4 | |
| 259217, 73442 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6030 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 1700063D12Rik; AI118035; AI840429; AW048913; AW556406; D5Mgi40; heat shock 70 kDa protein 12A; heat shock 70kD protein 12A; heat shock 70kDa protein 12A; Heat shock protein; heat shock protein 12A; heat shock protein family A (Hsp70) member 12A; HSP; HSPA12A; KIAA0417; mKIAA0417 | |
| A synthesized peptide derived from human HSPA12A (104-143AA). | |
| Primary | |
| -20°C | |
| HSPA12A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title